Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Menthyl Succinate 98.0+%, TCI America™
SDP

Catalog No. M248225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M248225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M248225G Supplier TCI America Supplier No. M248225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 77341-67-4
Molecular Formula C14H24O4
Molecular Weight (g/mol) 256.342
MDL Number MFCD23135646
InChI Key BLILOGGUTRWFNI-GRYCIOLGSA-N
Synonym Succinic Acid (-)-Menthyl Ester, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl Succinate, Succinic Acid (1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl Ester
PubChem CID 11219061
IUPAC Name 4-[(1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexyl]oxy-4-oxobutanoic acid
SMILES CC1CCC(C(C1)OC(=O)CCC(=O)O)C(C)C

Specifications

Melting Point 64°C
Color White
Quantity 25 g
Formula Weight 256.34
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (-)-Menthyl Succinate
EINECSNumber (3)-3281
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.