Learn More
Medchemexpress LLC MeO-suc-arg-pro-tyr-pNA | 82564-18-9 | 98.3% | 668.70 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP537610MG
Encompass_Preferred
MeO-Suc-Arg-Pro-Tyr-pNA is a biologically active peptide, also known as S-2586, that serves as a chymotrypsin substrate.
  • Biologically active peptide
  • Chymotrypsin substrate
  • Peptide and derivative
  • Enzyme substrate
Catalog No. 50-003-99695