Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MeO-Suc-Arg-Pro-Tyr-pNA | 82564-18-9 | 98.30% | 668.70 | 5 MG
SDP

Catalog No. 5000399111
Encompass_Preferred

MeOSuc-Arg-Pro-Tyr-pNA (S-2586) is a biologically active peptide that functions as a chymotrypsin substrate. It is intended for research use only. The compound appears as a white to off-white solid with the chemical formula C31H40N8O9. It is soluble in DMSO at 100 mg/mL.

  • Functions as a chymotrypsin substrate
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL
  • Chemical formula: C31H40N8O9
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-99111 Supplier Medchemexpress LLC Supplier No. HYP53765MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MeOSuc-Arg-Pro-Tyr-pNA (S-2586) is a biologically active peptide that functions as a chymotrypsin substrate. It is intended for research use only. The compound appears as a white to off-white solid with the chemical formula C31H40N8O9. It is soluble in DMSO at 100 mg/mL.

  • Functions as a chymotrypsin substrate
  • Appears as a white to off-white solid
  • Soluble in DMSO at 100 mg/mL
  • Chemical formula: C31H40N8O9
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.