Learn More
Medchemexpress LLC Meosuc-aapv-amc | 72252-90-5 | MFCD00077135 | 98.1% | 627.69 g/mol | C31H41N5O9 | 5 MG
SDP
Catalog No. 5000221857
Encompass_Preferred
MeOSuc-AAPV-AMC is a fluorogenic peptide substrate that releases 7-amino-4-methylcoumarin (AMC) upon proteolytic cleavage, producing a measurable fluorescence signal for elastase activity assays (Ex 380 nm, Em 460 nm). Intended for research use in enzymatic and kinetic studies.
  • Fluorogenic substrate for human leukocyte and porcine pancreatic elastase.
  • Generates AMC fluorescence at Ex 380 nm and Em 460 nm.
  • Reported Km for elastase: 362 μM.
  • High purity (98.07%) suitable for analytical assays.
  • Soluble in DMSO at 100 mg/mL; ultrasonication recommended.
  • White to off-white solid; powder storage at low temperature preserves stability.
  • Provided in milligram quantities for assay development and validation.
Catalog No. 50-002-21857 Supplier Medchemexpress LLC Supplier No. HY1368655MG
MeOSuc-AAPV-AMC is a fluorogenic peptide substrate that releases 7-amino-4-methylcoumarin (AMC) upon proteolytic cleavage, producing a measurable fluorescence signal for elastase activity assays (Ex 380 nm, Em 460 nm). Intended for research use in enzymatic and kinetic studies.
  • Fluorogenic substrate for human leukocyte and porcine pancreatic elastase.
  • Generates AMC fluorescence at Ex 380 nm and Em 460 nm.
  • Reported Km for elastase: 362 μM.
  • High purity (98.07%) suitable for analytical assays.
  • Soluble in DMSO at 100 mg/mL; ultrasonication recommended.
  • White to off-white solid; powder storage at low temperature preserves stability.
  • Provided in milligram quantities for assay development and validation.