Learn More
Enzo Life Sciences MeOSuc-AAPV-AMC (5 mg) CAS: 72252-90-5
SDP
Catalog No. NC0815207
Encompass_Preferred
Elastase substrate. Highly sensitive substrate for neutrophil elastase (leukocyte elastase) (Km=130 µM), but not granzyme H. This substrate can be used for in vitro assays with purified enzyme, on live cells, or in conditioned medium. Ex: 380 nm, Em: 460 nm. Alternative Name: Neutrophil elastase substrate (fluorogenic). Formula: C31H41N5O9. MW: 627.7. CAS: 72252-90-5. Purity: ≥95% (HPLC). Sequence N-Methoxysuccinyl-Ala-Ala-Pro-Val-7-amino-4-methylcoumarin. Appearance: White solid. Long Term Storage: -20°C
Catalog No. NC0815207 Supplier Enzo Life Sciences Supplier No. BMLP2240005
Elastase substrate. Highly sensitive substrate for neutrophil elastase (leukocyte elastase) (Km=130 µM), but not granzyme H. This substrate can be used for in vitro assays with purified enzyme, on live cells, or in conditioned medium. Ex: 380 nm, Em: 460 nm. Alternative Name: Neutrophil elastase substrate (fluorogenic). Formula: C31H41N5O9. MW: 627.7. CAS: 72252-90-5. Purity: ≥95% (HPLC). Sequence N-Methoxysuccinyl-Ala-Ala-Pro-Val-7-amino-4-methylcoumarin. Appearance: White solid. Long Term Storage: -20°C