Learn More
Cayman Chemical MeOSuc-AAPV-pNA, Purity: ≥98%, CAS: 70967-90-7, 100 mg
SDP
Catalog No. 502967455
Encompass_Preferred
A highly sensitive peptide substrate for both human and mouse neutrophil elastase and PR3
Catalog No. 50-296-7455 Supplier Cayman Chemical Supplier No. 17601100 MG
A highly sensitive peptide substrate for both human and mouse neutrophil elastase and PR3