Learn More
Enzo Life Sciences MeOSuc-AAPV-pNA (5mg). CAS: 70967-90-7
SDP
Catalog No. 502011288
Encompass_Preferred
Highly sensitive substrate for human and mouse neutrophil elastase (leukocyte elastase) and neutrophil proteinase 3 (PR-3, myeloblastin), but not cathepsin G nor chymotrypsin. This substrate can be used for in vitro assays with purified enzyme, or with biological fluids or conditioned medium. Hydrolysis of the substrate can be detected at 405-410 nm. Purity: ≥98%. Solubility: Soluble in DMSO (>20 mM) or methanol (1 mg/mL). Long Term Storage: -20°C.
Catalog No. 50-201-1288 Supplier Enzo Life Sciences Supplier No. BMLP2130005
Highly sensitive substrate for human and mouse neutrophil elastase (leukocyte elastase) and neutrophil proteinase 3 (PR-3, myeloblastin), but not cathepsin G nor chymotrypsin. This substrate can be used for in vitro assays with purified enzyme, or with biological fluids or conditioned medium. Hydrolysis of the substrate can be detected at 405-410 nm. Purity: ≥98%. Solubility: Soluble in DMSO (>20 mM) or methanol (1 mg/mL). Long Term Storage: -20°C.