Learn More
Bachem MeOSuc-Ala-Phe-Lys-AMC
SDP
Catalog No. 501946935
Encompass_Preferred
MeOSuc-Ala-Phe-Lys-AMC Trifluoroacetate, 201853-92-1, C33H41N5O8, Mr 635.72, 250mg. MeOSuc-AFK-AMC, a highly sensitive fluorogenic substrate for plasmin.
Catalog No. 50-194-6935 Supplier Bachem Supplier No. 4006754.0250
MeOSuc-Ala-Phe-Lys-AMC Trifluoroacetate, 201853-92-1, C33H41N5O8, Mr 635.72, 250mg. MeOSuc-AFK-AMC, a highly sensitive fluorogenic substrate for plasmin.