Learn More
Cayman Chemical MeOSuc-Ala-Ala-Pro-Val-AMC, Purity: ≥98%, CAS: 72252-90-5, 5 mg
SDP
Catalog No. 502970070
Encompass_Preferred
A fluorogenic substrate for human leukocyte and porcine pancreatic elastase; excitation: 355-380 nm; emission 440-460 nm
Catalog No. 50-297-0070 Supplier Cayman Chemical Supplier No. 149075 MG
A fluorogenic substrate for human leukocyte and porcine pancreatic elastase; excitation: 355-380 nm; emission 440-460 nm