Learn More
Bachem MeOSuc-Ala-Ala-Pro-Val-AMC
SDP
Catalog No. NC1766503
Encompass_Preferred
MeOSuc-Ala-Ala-Pro-Val-AMC, 72252-90-5, C31H41N5O9, Mr 627.70, 50mg. A highly sensitive fluorogenic substrate for the assay of human neutrophil and porcine pancreatic elastases. MeOSuc-AAPV-AMC is also cleaved by myeloblastin.
Catalog No. NC1766503 Supplier Bachem Supplier No. 4005227.0050
Add to Cart
Add to Cart
MeOSuc-Ala-Ala-Pro-Val-AMC, 72252-90-5, C31H41N5O9, Mr 627.70, 50mg. A highly sensitive fluorogenic substrate for the assay of human neutrophil and porcine pancreatic elastases. MeOSuc-AAPV-AMC is also cleaved by myeloblastin.