Learn More
Bachem MeOSuc-Ala-Ala-Pro-Val-pNA
SDP
Catalog No. 50260400
Encompass_Preferred
MeOSuc-Ala-Ala-Pro-Val-pNA, 70967-90-7, C27H38N6O9, Mr 590.63, 50mg. An excellent, highly soluble and specific substrate for a routine assay of human and rat leukocyte and porcine pancreatic elastases. MeOSuc-AAPV-pNA is also cleaved by myeloblastin.
Catalog No. 50-260-400 Supplier Bachem Supplier No. 4004221.0050
Add to Cart
Add to Cart
MeOSuc-Ala-Ala-Pro-Val-pNA, 70967-90-7, C27H38N6O9, Mr 590.63, 50mg. An excellent, highly soluble and specific substrate for a routine assay of human and rat leukocyte and porcine pancreatic elastases. MeOSuc-AAPV-pNA is also cleaved by myeloblastin.