Learn More
Medchemexpress LLC MeOSuc-Ala-Ala-Pro-Val-pNA | 70967-90-7 | 98.13% | 590.63 | 10 MG
SDP
Catalog No. 5000397342
Encompass_Preferred
MeOSuc-Ala-Ala-Pro-Val-pNA is a sensitive chromogenic substrate for human leukocyte and porcine pancreatic elastase. It is intended for research use only.
  • Exhibits kcat/Km values of 185,000, 15,000, 58,000 and 330,000 M⁻¹s⁻¹.
  • Active at 0.10 M phosphate, 0.20 M Mes, 0.10 M phosphate and 0.04 M phosphate for human leukocyte elastase reactions.
Catalog No. 50-003-97342 Supplier Medchemexpress LLC Supplier No. HYP268510MG
MeOSuc-Ala-Ala-Pro-Val-pNA is a sensitive chromogenic substrate for human leukocyte and porcine pancreatic elastase. It is intended for research use only.
  • Exhibits kcat/Km values of 185,000, 15,000, 58,000 and 330,000 M⁻¹s⁻¹.
  • Active at 0.10 M phosphate, 0.20 M Mes, 0.10 M phosphate and 0.04 M phosphate for human leukocyte elastase reactions.