Learn More
Cayman Chemical MeO-Suc-RPY-pNA (trifluoroacetate salt), Purity: ≥95%, 10 mg
SDP
Catalog No. 502940004
Encompass_Preferred
A colorimetric serine protease substrate; cleavage of the RPY peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of serine protease activity
Catalog No. 50-294-0004 Supplier Cayman Chemical Supplier No. 2770610 MG
A colorimetric serine protease substrate; cleavage of the RPY peptide sequence releases pNA, which can be quantified by colorimetric detection at 405 nm as a measure of serine protease activity