Learn More
Aviva Systems Biology Corp MEOX1 Antibody - N-terminal region (ARP32697_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32697_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MEOX1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MEOX1. Alternate names: MOX1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54712