Learn More
Echelon Biosciences Research Labs 2-C-Methyl-D-erythritol 4-phosphate (MEP) 10 mg
SDP
Supplier:  Echelon Biosciences Research Labs IM05110MG
Encompass
MEP is formed from 1-deoxy-D-xylulose 5-phosphate (DXP) by MEP Synthase (DXP reductoisomerase) and coupled with CMP to form 4-diphosphocytidyl-2-C-methyl-D-erythritol (CDP-ME) by CDPME Synthase. This item is non returnable
Catalog No. 50-720-400