Learn More
TARGETMOL CHEMICALS INC MEPACRINE 5MG
SDP
Catalog No. 502580806
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mepacrine (Erion) is an acridine derivative formerly widely used as an antimalarial. It is used in cell biological experiments as an inhibitor of phospholipase A2. purity: 100%
Catalog No. 50-258-0806 Supplier TARGETMOL CHEMICALS INC Supplier No. T244495MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mepacrine (Erion) is an acridine derivative formerly widely used as an antimalarial. It is used in cell biological experiments as an inhibitor of phospholipase A2. purity: 100%