Learn More
FUJIFILM Biosciences Mepanipyrim Standard
Catalog No. 5000450430
Encompass_Preferred
Mepanipyrim Standard is intended for pesticide residue analysis particularly in quality control and research settings With dual assay methods HPLC and qNMR the compound ensures reliable performance in analytical laboratories The product is packaged in glass to maintain stability and purity and must be stored at 2 10C protected from light
Catalog No. 50-004-50430 Supplier FUJIFILM Biosciences Supplier No. 13712651
Mepanipyrim Standard is intended for pesticide residue analysis particularly in quality control and research settings With dual assay methods HPLC and qNMR the compound ensures reliable performance in analytical laboratories The product is packaged in glass to maintain stability and purity and must be stored at 2 10C protected from light