Learn More
TARGETMOL CHEMICALS INC MEPAZINE 50MG
SDP
Catalog No. 502581193
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mepazine (Pecazine) is a potent and selective inhibitor of MALT1. Mepazine inhibits GSTMALT1 full length and GSTMALT1 325-760 with IC50s of 0.83 uM and 0.42 uM. Mepazine enhances apoptosis and affects the cell viability. purity: 99%
Catalog No. 50-258-1193 Supplier TARGETMOL CHEMICALS INC Supplier No. T1604050MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mepazine (Pecazine) is a potent and selective inhibitor of MALT1. Mepazine inhibits GSTMALT1 full length and GSTMALT1 325-760 with IC50s of 0.83 uM and 0.42 uM. Mepazine enhances apoptosis and affects the cell viability. purity: 99%