Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mepazine hydrochloride | 2975-36-2 | 99.3% | 10 MG
SDP

Catalog No. 5000232499
Encompass_Preferred

Mepazine hydrochloride is a small-molecule MALT1 protease inhibitor used in cellular and biochemical research to study apoptosis and signaling pathways.

  • High listed purity (99.25%).
  • Molecular weight 346.92 g/mol and formula C19H23ClN2S.
  • Available in small research pack sizes including 10 MG.
  • Suitable for cellular assays probing MALT1 activity and apoptosis.
  • Storage: solid at 4°C; solutions at -80°C for long-term, sealed and protected from moisture.

Catalog No. 50-002-32499 Supplier Medchemexpress LLC Supplier No. HY121282A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mepazine hydrochloride is a small-molecule MALT1 protease inhibitor used in cellular and biochemical research to study apoptosis and signaling pathways.

  • High listed purity (99.25%).
  • Molecular weight 346.92 g/mol and formula C19H23ClN2S.
  • Available in small research pack sizes including 10 MG.
  • Suitable for cellular assays probing MALT1 activity and apoptosis.
  • Storage: solid at 4°C; solutions at -80°C for long-term, sealed and protected from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.