Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mepazine hydrochloride | 2975-36-2 | 99.3% | 5 MG
SDP

Catalog No. 5000234815
Encompass_Preferred

Mepazine hydrochloride is the hydrochloride salt of mepazine, supplied for research use as a solid or as a DMSO solution and intended for in vitro biochemical and cellular assays.

  • High purity (99.25%).
  • Molecular formula C19H23ClN2S.
  • Molecular weight 346.92 g/mol.
  • Available in small solid packs and a 10 mM DMSO solution format.
  • Store sealed at 4°C; in solution, store at -80°C for long-term stability.

Catalog No. 50-002-34815 Supplier Medchemexpress LLC Supplier No. HY121282A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mepazine hydrochloride is the hydrochloride salt of mepazine, supplied for research use as a solid or as a DMSO solution and intended for in vitro biochemical and cellular assays.

  • High purity (99.25%).
  • Molecular formula C19H23ClN2S.
  • Molecular weight 346.92 g/mol.
  • Available in small solid packs and a 10 mM DMSO solution format.
  • Store sealed at 4°C; in solution, store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.