Learn More
Apexbio Technology LLC Mepenzolate Bromide 76-90-4 1g
SDP
Catalog No. 5000095149
Encompass_Preferred
Mepenzolate bromide (CAS 76-90-4) is a synthetic quaternary ammonium compound that functions as a muscarinic acetylcholine receptor antagonist By inhibiting parasympathetic neural transmission it reduces gastric secretion and gastrointestinal motility Mepenzolate bromide is generally supplied as a white to light cream powder with high solubility in methanol limited solubility in water and chloroform and minimal solubility in ether This compound is widely employed in research investigating modulation of cholinergic signaling gastrointestinal physiology and autonomic pharmacology and serves as a tool compound for studying antimuscarinic mechanisms in vitro and in vivo
Catalog No. 50-000-95149 Supplier Apexbio Technology LLC Supplier No. A84671000
Mepenzolate bromide (CAS 76-90-4) is a synthetic quaternary ammonium compound that functions as a muscarinic acetylcholine receptor antagonist By inhibiting parasympathetic neural transmission it reduces gastric secretion and gastrointestinal motility Mepenzolate bromide is generally supplied as a white to light cream powder with high solubility in methanol limited solubility in water and chloroform and minimal solubility in ether This compound is widely employed in research investigating modulation of cholinergic signaling gastrointestinal physiology and autonomic pharmacology and serves as a tool compound for studying antimuscarinic mechanisms in vitro and in vivo