Learn More
Medchemexpress LLC Mepenzolate bromide | 76-90-4 | 99.9% | 420.34 | 100 MG
SDP
Catalog No. 5000387470
Encompass_Preferred
Mepenzolate bromide is an orally administered muscarinic receptor antagonist with Kis of 0.68 and 2.6 nM for hM2R and hM3R, respectively. It can be used to suppress gastrointestinal hypermotility associated with irritable bowel syndrome. Mepenzolate bromide is also a GPR109A inhibitor.
  • Muscarinic receptor antagonist
  • GPR109A inhibitor
  • Exerts anti-inflammatory effect via a muscarinic receptor-independent mechanism
  • Bronchodilatory effect via a muscarinic receptor-dependent mechanism
  • Subtype-non-specific muscarinic antagonist
  • Beneficial effects in a mouse model of chronic obstructive pulmonary disease (COPD)
Catalog No. 50-003-87470 Supplier Medchemexpress LLC Supplier No. HY17585100MG
Mepenzolate bromide is an orally administered muscarinic receptor antagonist with Kis of 0.68 and 2.6 nM for hM2R and hM3R, respectively. It can be used to suppress gastrointestinal hypermotility associated with irritable bowel syndrome. Mepenzolate bromide is also a GPR109A inhibitor.
  • Muscarinic receptor antagonist
  • GPR109A inhibitor
  • Exerts anti-inflammatory effect via a muscarinic receptor-independent mechanism
  • Bronchodilatory effect via a muscarinic receptor-dependent mechanism
  • Subtype-non-specific muscarinic antagonist
  • Beneficial effects in a mouse model of chronic obstructive pulmonary disease (COPD)