Learn More
Cayman Chemical Mepenzolate (bromide), Purity: ≥95%, CAS: 76-90-4, 500 mg
SDP
Catalog No. 502994935
Encompass_Preferred
A muscarinic M2 and M3 receptor antagonist (Kis = 0.68 and 2.6 nM, respectively); has antispasmodic activity in cats and dogs when administered at a dose of 0.5 mg/kg; decreases airspace enlargement induced by elastase or cigarette smoke in a mouse model of COPD; reduces oxidative stress, decreases the expression of pro-inflammatory cytokines, and increases the expression of anti-inflammatory cytokines in a mouse model of diabetic wound healing
Catalog No. 50-299-4935 Supplier Cayman Chemical Supplier No. 23717500 MG
A muscarinic M2 and M3 receptor antagonist (Kis = 0.68 and 2.6 nM, respectively); has antispasmodic activity in cats and dogs when administered at a dose of 0.5 mg/kg; decreases airspace enlargement induced by elastase or cigarette smoke in a mouse model of COPD; reduces oxidative stress, decreases the expression of pro-inflammatory cytokines, and increases the expression of anti-inflammatory cytokines in a mouse model of diabetic wound healing