Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mephenesin | 59-47-2 | 99.69% | 500 MG
SDP

Supplier:  Medchemexpress LLC HYB1283500MG

Encompass_Preferred

Mephenesin is an NMDA receptor antagonist and a central muscle relaxant with antianxiety, muscle-paralyzing, and anticonvulsant effects. It directly affects skeletal muscle fibers, leading to skeletal muscle relaxation and is being researched for its potential in treating spasticity or painful muscle spasms. This product is provided as a white to off-white solid.

  • Molecular weight: 182.22
  • Formula: C10H14O3
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 99.69%

Catalog No. 50-004-11834


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.