Learn More
TARGETMOL CHEMICALS INC Mephenytoin 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T2142510MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Mephenytoin (Phenantoin) is an anticonvulsant and a substrate of CYP2C19 and CYP2B6. Purity 99.72%
Catalog No. 50-287-9615