Learn More
Medchemexpress LLC Mephenytoin | 50-12-4 | >98% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB118450MG
Encompass_Preferred
Mephenytoin is an anticonvulsant and a substrate of the cytochrome P450 (CYP) isoform CYP2C19. It has been utilized in the analysis of cytochrome P450 metabolism. This compound is associated with central nervous system (CNS) adverse effects and potentially life-threatening idiosyncratic reactions, including skin rash, fever, and blood dyscrasias.
  • Anticonvulsant
  • CYP2C19 and CYP2B6 substrate
  • Used in analysis of cytochrome P450 metabolism
Catalog No. 50-004-12604