Learn More
Medchemexpress LLC Mephenytoin | 50-12-4 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB118410MM/1ML
Encompass_Preferred
Mephenytoin is an anticonvulsant and a substrate for CYP2C19 and CYP2B6. It is for research use only and not sold to patients.
  • Anticonvulsant activity
  • Substrate for CYP2C19 and CYP2B6
  • Reduces maternal weight gain and increases offspring mortality at 200 mg/kg in pregnant Sprague-Dawley CD rats.
  • Significantly reduces serum cholesterol and triglyceride levels in mice at 20 mg/kg per day for 16 days, suggesting a hypolipidemic effect.
Catalog No. 50-004-13609