Learn More
Medchemexpress LLC Mephenytoin | 50-12-4 | 99.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB11845MG
Encompass_Preferred
Mephenytoin is an anticonvulsant and a substrate for CYP2C19 and CYP2B6. It is for research use only. In preclinical studies, orally administered Mephenytoin reduced maternal weight gain and increased offspring mortality at 200 mg/kg in pregnant Sprague-Dawley CD rats. Additionally, Mephenytoin significantly reduced serum cholesterol and triglyceride levels in mice, suggesting a hypolipidemic effect.
  • Anticonvulsant compound.
  • Acts as a substrate for CYP2C19 and CYP2B6.
  • Reduced maternal weight gain in preclinical studies at higher doses.
  • Demonstrated hypolipidemic effect in mice by reducing serum cholesterol and triglyceride levels.
  • Intended for research use only.
Catalog No. 50-004-12221