Learn More
Medchemexpress LLC Mephenytoin | 50-12-4 | MFCD00022407 | 100.0% | 218.26 g/mol | C12H14N2O2 | 10 MG
SDP
Catalog No. 5000297989
Encompass_Preferred
Mephenytoin is an anticonvulsant compound supplied for research and analytical use. It is commonly used as a probe substrate for cytochrome P450 enzymes (notably CYP2C19 and CYP2B6) in phenotyping and metabolic studies, and is provided as a high-purity solid suitable for laboratory assays.
  • Anticonvulsant compound used in research applications.
  • Substrate for CYP2C19 and CYP2B6 for metabolism studies.
  • High purity, suitable for analytical work and assays.
  • White to off-white solid appearance for easy handling.
  • Available in small research quantities and as solution formulations.
Catalog No. 50-002-97989 Supplier Medchemexpress LLC Supplier No. HYB118410MG
Mephenytoin is an anticonvulsant compound supplied for research and analytical use. It is commonly used as a probe substrate for cytochrome P450 enzymes (notably CYP2C19 and CYP2B6) in phenotyping and metabolic studies, and is provided as a high-purity solid suitable for laboratory assays.
  • Anticonvulsant compound used in research applications.
  • Substrate for CYP2C19 and CYP2B6 for metabolism studies.
  • High purity, suitable for analytical work and assays.
  • White to off-white solid appearance for easy handling.
  • Available in small research quantities and as solution formulations.