Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (+)-Mephenytoin | 70989-04-7 | 99.9% | 1 ML
SDP

Catalog No. 5000413610
Encompass_Preferred

(S)-Mephenytoin is an anticonvulsive agent that functions as a substrate of the cytochrome P450 (CYP) isoform CYP2C19. It is utilized for the analysis of cytochrome P450 metabolism.

  • Functions as an anticonvulsive agent
  • Substrate of the cytochrome P450 (CYP) isoform CYP2C19
  • Used for the analysis of cytochrome P450 metabolism
  • Supplied as a 10 mM solution in 1 mL of DMSO
  • High purity of 99.94%
  • Molecular weight of 218.25
  • Molecular formula C12H14N2O2

Catalog No. 50-004-13610 Supplier Medchemexpress LLC Supplier No. HYB1184A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Mephenytoin is an anticonvulsive agent that functions as a substrate of the cytochrome P450 (CYP) isoform CYP2C19. It is utilized for the analysis of cytochrome P450 metabolism.

  • Functions as an anticonvulsive agent
  • Substrate of the cytochrome P450 (CYP) isoform CYP2C19
  • Used for the analysis of cytochrome P450 metabolism
  • Supplied as a 10 mM solution in 1 mL of DMSO
  • High purity of 99.94%
  • Molecular weight of 218.25
  • Molecular formula C12H14N2O2

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.