Learn More
Medchemexpress LLC Mephenytoin-d5 | 1185032-66-9 | >98.72% | 1 MG
SDP
Catalog No. 5000413820
Encompass_Preferred
Mephenytoin-d5 is a deuterium labeled form of Mephenytoin, an anticonvulsant that acts as a substrate for CYP2C19 and CYP2B6. It is commonly utilized as a tracer and internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer for quantitative analysis
  • Can be used as an internal standard for quantitative analysis
  • Deuteration may affect pharmacokinetic and metabolic profiles
Catalog No. 50-004-13820 Supplier Medchemexpress LLC Supplier No. HYB1184S11MG
Mephenytoin-d5 is a deuterium labeled form of Mephenytoin, an anticonvulsant that acts as a substrate for CYP2C19 and CYP2B6. It is commonly utilized as a tracer and internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer for quantitative analysis
  • Can be used as an internal standard for quantitative analysis
  • Deuteration may affect pharmacokinetic and metabolic profiles