Learn More
Medchemexpress LLC MePhos | 251320-86-2 | 98.0% | 364.50 | 25 G
SDP
Catalog No. 5000367087
Encompass_Preferred
MePhos is an o-BPP ligand. The combination of MePhos and silver carbonate serves as a catalyst for H/D exchange reactions.
  • Designed as an o-BPP ligand
  • Can be used with silver carbonate as a catalyst
  • Facilitates H/D exchange reactions
Catalog No. 50-003-67087 Supplier Medchemexpress LLC Supplier No. 25GHYW009062
MePhos is an o-BPP ligand. The combination of MePhos and silver carbonate serves as a catalyst for H/D exchange reactions.
  • Designed as an o-BPP ligand
  • Can be used with silver carbonate as a catalyst
  • Facilitates H/D exchange reactions