Learn More
Medchemexpress LLC Mepiquat chloride | 24307-26-4 | MFCD00055543 | ≥97.0% | 149.66 g/mol | C7H16ClN | 100 G
SDP
Catalog No. 5000300184
Encompass_Preferred
Mepiquat chloride (N,N-Dimethylpiperidinium chloride; PIX) is a quaternary ammonium salt used as a plant growth regulator to reduce vegetative growth and plant height. Supplied as a solid reagent for research and analytical use, it is suitable for plant physiology studies, growth regulation assays, and use as a reference standard.
  • Potent plant growth regulator that decreases plant height.
  • High purity solid reagent (≥97.0%).
  • Molecular formula C7H16ClN and molecular weight 149.66 g/mol.
  • Available in multiple pack sizes, including 100 g.
  • Suitable for research, analytical standards, and physiological assays.
Catalog No. 50-003-00184 Supplier Medchemexpress LLC Supplier No. HYW017448100G
Mepiquat chloride (N,N-Dimethylpiperidinium chloride; PIX) is a quaternary ammonium salt used as a plant growth regulator to reduce vegetative growth and plant height. Supplied as a solid reagent for research and analytical use, it is suitable for plant physiology studies, growth regulation assays, and use as a reference standard.
  • Potent plant growth regulator that decreases plant height.
  • High purity solid reagent (≥97.0%).
  • Molecular formula C7H16ClN and molecular weight 149.66 g/mol.
  • Available in multiple pack sizes, including 100 g.
  • Suitable for research, analytical standards, and physiological assays.