Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mepiquat chloride (N,N-dimethylpiperidinium chloride) | 24307-26-4 | MFCD00055543 | >97.0% | 149.66 | C7H16ClN | 25 G
SDP

Catalog No. 5000303589
Encompass_Preferred

Mepiquat chloride is a quaternary ammonium salt used as a plant growth regulator and analytical standard for research applications. It reduces plant height and is supplied as a white to off-white solid with high purity and defined storage recommendations for ambient and frozen conditions.

  • Used as a plant growth regulator and analytical standard for research.
  • Molecular formula C7H16ClN; molecular weight 149.66.
  • Purity ≥97.0%.
  • Appearance white to off-white solid.
  • Storage: room temperature for 3 years; in solvent -80°C for 2 years, -20°C for 1 year.
  • Available in 25 g, 50 g, 100 g, and larger quantities by quote.

Catalog No. 50-003-03589 Supplier Medchemexpress LLC Supplier No. 25GHYW017448
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mepiquat chloride is a quaternary ammonium salt used as a plant growth regulator and analytical standard for research applications. It reduces plant height and is supplied as a white to off-white solid with high purity and defined storage recommendations for ambient and frozen conditions.

  • Used as a plant growth regulator and analytical standard for research.
  • Molecular formula C7H16ClN; molecular weight 149.66.
  • Purity ≥97.0%.
  • Appearance white to off-white solid.
  • Storage: room temperature for 3 years; in solvent -80°C for 2 years, -20°C for 1 year.
  • Available in 25 g, 50 g, 100 g, and larger quantities by quote.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.