Learn More
Cayman Chemical Mepivacaine-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1346597-90-7, 1 mg
SDP
Catalog No. 502978523
Encompass_Preferred
An internal standard for the quantification of mepivacaine by GC- or LC-MS
Catalog No. 50-297-8523 Supplier Cayman Chemical Supplier No. 398121 MG
An internal standard for the quantification of mepivacaine by GC- or LC-MS