Learn More
Medchemexpress LLC (+)-Mepivacaine | 24358-84-7 | 99.7% | 10 MG
SDP
Catalog No. 5000233324
Encompass_Preferred
(+)-Mepivacaine is the (+) enantiomer of mepivacaine, an amide-type local anesthetic that blocks voltage-gated sodium channels to produce temporary loss of sensation. Supplied as a high-purity research chemical for pharmacological and electrophysiological studies.
  • High purity (99.7%), suitable for research applications.
  • Acts as a sodium channel blocker for studies of neuronal excitability.
  • Available in small milligram quantities for laboratory use (5 mg, 10 mg, 25 mg).
  • Molecular formula C15H22N2O; molecular weight 246.35.
  • Offered as the (+) enantiomer for enantioselective research.
Catalog No. 50-002-33324 Supplier Medchemexpress LLC Supplier No. HY11996110MG
(+)-Mepivacaine is the (+) enantiomer of mepivacaine, an amide-type local anesthetic that blocks voltage-gated sodium channels to produce temporary loss of sensation. Supplied as a high-purity research chemical for pharmacological and electrophysiological studies.
  • High purity (99.7%), suitable for research applications.
  • Acts as a sodium channel blocker for studies of neuronal excitability.
  • Available in small milligram quantities for laboratory use (5 mg, 10 mg, 25 mg).
  • Molecular formula C15H22N2O; molecular weight 246.35.
  • Offered as the (+) enantiomer for enantioselective research.