Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (+)-Mepivacaine | 24358-84-7 | 99.7% | 5 MG
SDP

Catalog No. 5000233309
Encompass_Preferred

(+)-Mepivacaine is the optically active (+)-enantiomer of the local anesthetic mepivacaine supplied as a research-grade reagent for studies of sodium channel function, membrane physiology, and neuropharmacology.

  • High purity: 99.7%.
  • Chemical formula: C15H22N2O.
  • Molecular weight: 246.35 g/mol.
  • Pack size: 5 mg.
  • Recommended storage: -20°C (powder); in solvent -80°C for long-term storage.

Catalog No. 50-002-33309 Supplier Medchemexpress LLC Supplier No. HY1199615MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(+)-Mepivacaine is the optically active (+)-enantiomer of the local anesthetic mepivacaine supplied as a research-grade reagent for studies of sodium channel function, membrane physiology, and neuropharmacology.

  • High purity: 99.7%.
  • Chemical formula: C15H22N2O.
  • Molecular weight: 246.35 g/mol.
  • Pack size: 5 mg.
  • Recommended storage: -20°C (powder); in solvent -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.