Learn More
Apexbio Technology LLC Mepivacaine HCl 1722-62-9 50mg
SDP
Catalog No. 5000093013
Encompass_Preferred
Mepivacaine hydrochloride (CAS 1722-62-9) is a synthetic tertiary amine local anesthetic It exerts its effect by reversibly inhibiting voltage-gated sodium channels on neuronal cell membranes thereby blocking nerve impulse conduction This action results in loss of sensation in targeted tissues without affecting consciousness Mepivacaine is frequently utilized in research settings to investigate mechanisms of nerve blockade neuronal excitability and pain signal transmission and serves as a standard control in studies comparing the pharmacological profiles of local anesthetic agents
Catalog No. 50-000-93013 Supplier Apexbio Technology LLC Supplier No. B196750
Mepivacaine hydrochloride (CAS 1722-62-9) is a synthetic tertiary amine local anesthetic It exerts its effect by reversibly inhibiting voltage-gated sodium channels on neuronal cell membranes thereby blocking nerve impulse conduction This action results in loss of sensation in targeted tissues without affecting consciousness Mepivacaine is frequently utilized in research settings to investigate mechanisms of nerve blockade neuronal excitability and pain signal transmission and serves as a standard control in studies comparing the pharmacological profiles of local anesthetic agents