Learn More
AdipoGen Mepivacaine hydrochloride (50 mg)
SDP
Supplier:  AdipoGen CDXM0557M050
Encompass_Preferred
Chemical. CAS: 1722-62-9. Formula: C15H22N2O . HCl. MW: 282.81. Mepivacaine hydrochloride is a long-acting local anesthetic that reversibly blocks transient Na+ inward current, as well as the steady-state K+ outward current. Mepivacaine blocks tandem pore (TASK) and Kv1.5 potassium channels in model systems.
Catalog No. 50-205-4544