Learn More
Med Vet International Zoetis Carbocaine-V 2% Sterile Aqueous Solution, 50mL
Catalog No. 502828885
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
CARBOCAINE-V Sterile Aqueous Solution (2% mepivacaine hydrochloride, USP) is recommended for infiltration, nerve block, intra-articular and epidural anesthesia for horses. It has also been found useful for topical anesthesia of the laryngeal mucosa prior to ventriculectomy.
Catalog No. 50-282-8885 Supplier Med Vet International Supplier No. RXCARBOCAINEV
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
CARBOCAINE-V Sterile Aqueous Solution (2% mepivacaine hydrochloride, USP) is recommended for infiltration, nerve block, intra-articular and epidural anesthesia for horses. It has also been found useful for topical anesthesia of the laryngeal mucosa prior to ventriculectomy.