Learn More
Medchemexpress LLC Mepivacaine hydrochloride | 1722-62-9 | 99.68% | 1 G
SDP
Catalog No. 5000405981
Encompass_Preferred
Mepivacaine hydrochloride is a local anesthetic that inhibits nerve impulse initiation and conduction by binding to voltage-gated sodium ion channels in neuronal cell membranes. This action blocks sodium influx and membrane depolarization, leading to a reversible loss of sensation. It is characterized by a rapid onset and moderate duration of action compared to other local anesthetics.
  • Exhibits a more rapid onset than some other local anesthetics.
  • Provides a moderate duration of action.
  • Shows preferential use-dependent block of Na(v)1.8 channels.
  • For research use only.
Catalog No. 50-004-05981 Supplier Medchemexpress LLC Supplier No. HYB0517A1G
Add to Cart
Add to Cart
Mepivacaine hydrochloride is a local anesthetic that inhibits nerve impulse initiation and conduction by binding to voltage-gated sodium ion channels in neuronal cell membranes. This action blocks sodium influx and membrane depolarization, leading to a reversible loss of sensation. It is characterized by a rapid onset and moderate duration of action compared to other local anesthetics.
  • Exhibits a more rapid onset than some other local anesthetics.
  • Provides a moderate duration of action.
  • Shows preferential use-dependent block of Na(v)1.8 channels.
  • For research use only.