Learn More
Medchemexpress LLC Mepivacaine hydrochloride | 1722-62-9 | 99.7% | 1 ML
SDP
Catalog No. 5000408802
Encompass_Preferred
Mepivacaine hydrochloride binds to specific voltage-gated sodium ion channels in neuronal cell membranes, inhibiting both sodium influx and membrane depolarization. This blockage of nerve impulse initiation and conduction results in a reversible loss of sensation. It has a more rapid onset and moderate duration of action compared to other local anesthetics. It also displays a preferential use-dependent block of Na(v)1.8.
  • Inhibits sodium influx and membrane depolarization
  • Results in reversible loss of sensation
  • Rapid onset of action
  • Moderate duration of action
  • Displays preferential use-dependent block of Na(v)1.8
Catalog No. 50-004-08802 Supplier Medchemexpress LLC Supplier No. HYB0517A10MM/1ML
Mepivacaine hydrochloride binds to specific voltage-gated sodium ion channels in neuronal cell membranes, inhibiting both sodium influx and membrane depolarization. This blockage of nerve impulse initiation and conduction results in a reversible loss of sensation. It has a more rapid onset and moderate duration of action compared to other local anesthetics. It also displays a preferential use-dependent block of Na(v)1.8.
  • Inhibits sodium influx and membrane depolarization
  • Results in reversible loss of sensation
  • Rapid onset of action
  • Moderate duration of action
  • Displays preferential use-dependent block of Na(v)1.8