Learn More
Medchemexpress LLC Meprednisone | 1247-42-3 | 99.6% | 5 MG
SDP
Catalog No. 5000406067
Encompass_Preferred
Meprednisone is a glucocorticoid and a methylated derivative of Prednisone. It modulates the glucocorticoid receptor, is orally active, and is intended for research use only. It can be used in research for conditions such as rheumatoid arthritis, scleroderma, and hypothyroidism.
  • Modulates the glucocorticoid receptor
  • Orally active
  • Intended for research use only
  • Potential research applications include rheumatoid arthritis, scleroderma, and hypothyroidism
  • May have side effects on fetuses and pregnant women
Catalog No. 50-004-06067 Supplier Medchemexpress LLC Supplier No. HYB02435MG
Meprednisone is a glucocorticoid and a methylated derivative of Prednisone. It modulates the glucocorticoid receptor, is orally active, and is intended for research use only. It can be used in research for conditions such as rheumatoid arthritis, scleroderma, and hypothyroidism.
  • Modulates the glucocorticoid receptor
  • Orally active
  • Intended for research use only
  • Potential research applications include rheumatoid arthritis, scleroderma, and hypothyroidism
  • May have side effects on fetuses and pregnant women