Learn More
Thermo Scientific™ MEPS™ for LC: Thermo Scientific, CTC Analytics, HTA and Varian 8400 Systems
Catalog No. 60308407
Change view
Click to view available options
Description:
C18 MEPS for LC
Hypercarb MEPS for LC
Retain PEP MEPS for LC
Retain PEP,-CX,-AX,Hypercarb, C18
Retain-AX MEPS for LC
Retain-CX MEPS for LC
Silica MEPS for LC
Verify-AX MEPS for LC
Verify-CX MEPS for LC
Sorbent:
HyperSep C18 MEPS
HyperSep Hypercarb MEPS
HyperSep Retain AX MEPS
HyperSep Retain CX MEPS
HyperSep Retain PEP MEPS
HyperSep Retain PEP, Retain CX, Retain AX, Hypercarb, and C18
HyperSep Silica MEPS
HyperSep Verify AX MEPS
HyperSep Verify CX MEPS
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Sorbent
60-308-407 C18 MEPS for LC HyperSep C18 MEPS
60-308-401 Retain PEP MEPS for LC HyperSep Retain PEP MEPS
60-308-402 Retain-CX MEPS for LC HyperSep Retain CX MEPS
60-308-403 Retain-AX MEPS for LC HyperSep Retain AX MEPS
60-308-404 Hypercarb MEPS for LC HyperSep Hypercarb MEPS
60-308-405 Verify-CX MEPS for LC HyperSep Verify CX MEPS
60-308-406 Verify-AX MEPS for LC HyperSep Verify AX MEPS
60-308-408 Silica MEPS for LC HyperSep Silica MEPS
60-308-409 Retain PEP,-CX,-AX,Hypercarb, C18 HyperSep Retain PEP, Retain CX, Retain AX, Hypercarb, and C18
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 60-308-407 Supplier Thermo Scientific™ Supplier No. 60308407
Save hours in sample preparation, and go from extraction to injection in a single process with these 250μL syringes for LC applications using CTC Analytics.
Save hours in sample preparation, and go from extraction to injection in a single process with Thermo Scientific™ HyperSep™ MEPS™ (Micro Extraction by Packed Sorbent) technology.
Suitable for reversed phases, normal phases, mixed mode or ion exchange chemistries, Thermo Scientific HyperSep MEPS Syringes and Components for Analytics are available in a variety of SPE phase chemistries. The HyperSep MEPS Barrel Insert and Needle Assembly (BIN) contains the stationary phase, and is built into the syringe needle.
These HyperSep MEPS BINs are designed for use with Thermo Scientific, CTC Analytics, HTA and Varian 8400 LC Systems.
MEPS performs the same functions as SPE—the removal of interfering matrix components and the selective isolation and concentration of analytes—except that it miniaturizes conventional SPE packed bed devices from milliliter bed volumes to microliter volumes, and incorporates them right into the syringe.
Mechanically robust, the technique offers significant advantages over conventional SPE extraction for method development and other GC applications:
Significantly reduces the time to prepare and inject samples from hours to minutes—typically less than two minutes per sample
Eliminates all extra steps between sample preparation and sample injection; the technique is fully compatible with direct injection onto GC
Reduces buffer and solvent volume from milliliters to microliters
Reduces the required sample volume to as little as 3.6µL, making the techniques especially well-suited for on-line use with volume-limited samples
Can easily be automated compatible and is compatible with CTC and eVol platforms
TRUSTED_SUSTAINABILITY
Specifications
Description C18 MEPS for LC
For Use With (Application) Liquid Chromatography
For Use With (Equipment) Thermo Scientific, CTC Analytics, HTA, and Varian 8400 Systems Using 100 μL and 250 μL MEPS Syringes
Product Line HyperSep
Sorbent HyperSep C18 MEPS
Type MEPS Barrel Insert and Needle (BIN)