Learn More
Apexbio Technology LLC Meptazinol HCl 59263-76-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063459
Encompass_Preferred
Meptazinol HCl also chemically known as WY-22811 hydrochloride is a centrally acting analgesic agent characterized as a partial agonist at the 1 opioid receptor Structurally it belongs to the synthetic opioid family and interacts selectively with opioid receptors in the central nervous system to modulate pain transmission Its partial agonist properties allow for analgesic activity with generally reduced risk of respiratory depression compared to full opioid agonists making it a useful compound in pharmacological studies examining opioid receptor activation modulation and analgesic effects In biomedical research settings Meptazinol HCl is commonly employed to investigate pain management mechanisms opioid receptor signaling pathways receptor selectivity profiles and analgesia-related side effect mitigation
Catalog No. 50-000-63459 Supplier Apexbio Technology LLC Supplier No. B21105.1
Meptazinol HCl also chemically known as WY-22811 hydrochloride is a centrally acting analgesic agent characterized as a partial agonist at the 1 opioid receptor Structurally it belongs to the synthetic opioid family and interacts selectively with opioid receptors in the central nervous system to modulate pain transmission Its partial agonist properties allow for analgesic activity with generally reduced risk of respiratory depression compared to full opioid agonists making it a useful compound in pharmacological studies examining opioid receptor activation modulation and analgesic effects In biomedical research settings Meptazinol HCl is commonly employed to investigate pain management mechanisms opioid receptor signaling pathways receptor selectivity profiles and analgesia-related side effect mitigation