Learn More
AdipoGen Meptyldinocap
SDP
Supplier:  AdipoGen CDXM0311M025
Encompass_Preferred
Chemical. CAS 131-72-6. Formula C18H24N2O6. MW 364.4. Synthetic. Powdery mildew Erysiphe necator dinitrophenol fungicide. Stops cellular energy production. Interferes with fungal respiration by acting as an uncoupler of oxidative phosphorylation. Compound can be used as analytical reference material.
Catalog No. 50-256-0467