Learn More
Apexbio Technology LLC Mepyramine maleate 59-33-6 100mg
SDP
Catalog No. 502240774
Encompass_Preferred
Mepyramine maleate (CAS 59-33-6) is a selective antagonist of the histamine H1 receptor exhibiting dissociation constants of 0 8 nM for H1 5200 nM for H2 and over 3000 nM for H3 receptors By blocking H1 receptor-mediated signaling it modulates pathways involved in allergic responses Mepyramine maleate is commonly utilized in experimental models of allergic diseases to investigate histamine-related mechanisms and to delineate the role of H1 receptor activity in immunological and inflammatory processes
This item is not returnable. View return policy
Catalog No. 50-224-0774 Supplier Apexbio Technology LLC Supplier No. B6396100
Mepyramine maleate (CAS 59-33-6) is a selective antagonist of the histamine H1 receptor exhibiting dissociation constants of 0 8 nM for H1 5200 nM for H2 and over 3000 nM for H3 receptors By blocking H1 receptor-mediated signaling it modulates pathways involved in allergic responses Mepyramine maleate is commonly utilized in experimental models of allergic diseases to investigate histamine-related mechanisms and to delineate the role of H1 receptor activity in immunological and inflammatory processes