Learn More
Apexbio Technology LLC Mepyramine maleate 59-33-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092060
Encompass_Preferred
Mepyramine maleate (CAS 59-33-6) is a selective antagonist of the histamine H1 receptor exhibiting dissociation constants of 0 8 nM for H1 5200 nM for H2 and over 3000 nM for H3 receptors By blocking H1 receptor-mediated signaling it modulates pathways involved in allergic responses Mepyramine maleate is commonly utilized in experimental models of allergic diseases to investigate histamine-related mechanisms and to delineate the role of H1 receptor activity in immunological and inflammatory processes
Catalog No. 50-000-92060 Supplier Apexbio Technology LLC Supplier No. B63965.1
Mepyramine maleate (CAS 59-33-6) is a selective antagonist of the histamine H1 receptor exhibiting dissociation constants of 0 8 nM for H1 5200 nM for H2 and over 3000 nM for H3 receptors By blocking H1 receptor-mediated signaling it modulates pathways involved in allergic responses Mepyramine maleate is commonly utilized in experimental models of allergic diseases to investigate histamine-related mechanisms and to delineate the role of H1 receptor activity in immunological and inflammatory processes