Learn More
Abcam Mepyramine maleate, H1 inverse agonist, 100MG
Catalog No. 502707323
Encompass_Preferred
MW 401.46 g/mol, Purity >99%. Selective H₁ receptor inverse agonist (Ki = 0.4 nM). Blood-brain barrier permeable. Shows antihistaminergic action. Additionally, inhibits histamine-induced inositol phosphate production and intracellular calcium mobilization.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7323 Supplier Abcam Supplier No. AB120735100MG
MW 401.46 g/mol, Purity >99%. Selective H₁ receptor inverse agonist (Ki = 0.4 nM). Blood-brain barrier permeable. Shows antihistaminergic action. Additionally, inhibits histamine-induced inositol phosphate production and intracellular calcium mobilization.
The product is subject to the following: Abcam Restricted Use Statement