Learn More
Medchemexpress LLC Mepyramine maleate (pyrilamine maleate) | 59-33-6 | 99.9% | 1 ML
SDP
Catalog No. 5000412981
Encompass_Preferred
Mepyramine maleate is a first-generation antihistamine that acts as an antagonist of the histamine H1 receptor. It is characterized by specific binding affinities for H1, H2, and H3 receptors.
  • Antagonist of the histamine H1 receptor
  • Binds to H1 receptor with Kd of 0.8 nM
  • Exhibits Kds of 5200 nM for H2 and >3000 nM for H3 receptors
  • Decreases InsP levels in CHO-gpH1 cells
  • Reduces maximal response to ATP in CHO-gpH1 cells
  • Demonstrates analgesic effects in vivo at specific dosages
Catalog No. 50-004-12981 Supplier Medchemexpress LLC Supplier No. HYB128110MM/1ML
Mepyramine maleate is a first-generation antihistamine that acts as an antagonist of the histamine H1 receptor. It is characterized by specific binding affinities for H1, H2, and H3 receptors.
  • Antagonist of the histamine H1 receptor
  • Binds to H1 receptor with Kd of 0.8 nM
  • Exhibits Kds of 5200 nM for H2 and >3000 nM for H3 receptors
  • Decreases InsP levels in CHO-gpH1 cells
  • Reduces maximal response to ATP in CHO-gpH1 cells
  • Demonstrates analgesic effects in vivo at specific dosages